Learn More
Electron Microscopy Sciences Wehnelt Aperture 400 µm Hole
Supplier: Electron Microscopy Sciences 49040TA
Made from 0.004" tantalum foil. Replaceable tantalum apertures for some microscopes (Amray and Philips). They improve the performance of the electron gun.