Learn More
Sklar Instruments Econo Webster Needle Holder, 5-1/4 in., Blunt, Straight, Smooth, Stainless Steel. BX12.
SDP
Supplier:  Sklar Instruments 21809
Encompass_Preferred
Econo Webster Needle Holder, 5-1/4 in., Blunt, Straight, Smooth, Stainless Steel. Econo Webster Needle Holder is used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth tips and a length of 5-1/4 inches. They are sold in boxes of 12.
Catalog No. 50-110-9758