Learn More
Tocris Bioscience™ WAY 316606 hydrochloride
Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling
$343.36 - $1438.40
Specifications
Purity | >99% |
---|---|
CAS | 1781835-02-6 |
Product Type | WAY 316606 hydrochloride |
Molecular Weight (g/mol) | 484.94 |
Inhibitors | WAY 316606 hydrochloride |
Description
Secreted Frizzled-Related Protein-1 (sFRP-1) inhibitor (IC50 = 0.65 μM); prevents sFRP-1 from interacting with Wnt and thus increases Wnt signaling. Increases total bone area in a murine calvarial organ culture assay.We are aware of a study in PLOS Biology where WAY-316606 was found to stimulate hair follicle growth. Please note that WAY-316606 from Tocris is for laboratory research use only, and that we will only sell to established scientific businesses and institutes.
Specifications
>99% | |
WAY 316606 hydrochloride | |
WAY 316606 hydrochloride |
1781835-02-6 | |
484.94 |
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.