Learn More
Electron Microscopy Sciences Wax Stripper 1 LB
SDP
Supplier:  Electron Microscopy Sciences 5039201
Encompass_Preferred
WaxStrip is an environmentally safe powder compound prepared primarily for the removal of mounting waxes in semiconductor processing. WaxStrip is compatible for use on silicon, glass, iron, aluminum, zinc, brass, copper, magnesium and other materials.
Catalog No. 50-997-393