Learn More
OHAUS™ Waving Incubating Shaker
OHAUS Incubating Waving Shakers are designed to incubate samples from 5° above ambient to 65°C to provide accurate and repeatable results.
Supplier: OHAUS™ 30391987/EMD
Description
- Waving shakers offer a smooth, low-foaming three-dimensional wave motion
- LED displays with touchpad controls for speed, tilt angle and time
- Electronic tilt adjustment from 0 to 20° while unit is operating
- Timer with audible alarm
Specifications
0 to 20° | |
110 V | |
5°C to 65°C | |
3.74 A | |
50/60 Hz | |
27.4 cm | |
41.9 cm | |
10.1in. | |
25.7 cm | |
14.5 lb. | |
ISWV02 HDG | |
3 wire cord and plug |
1 to 30 rpm | |
120 V | |
420 W | |
5 lb. | |
10.8in. | |
9.25in. | |
23.5 cm | |
7.25in. | |
18.4 cm | |
6.6 kg | |
80% | |
1 sec. to 160 hr. |