Learn More
Microtech Scientific Sterile 1000mL Coliform Bottle 64/case
Supplier: Microtech Scientific MS5RT1000PE
Sterile sample bottle used for the detection of coliforms and other microorganisms. Sample bottles conform to EPA standards for sterility, volume, graduation accuracy, and non-autoflourescence.