Learn More
Cole-Parmer Stuart D4000/Euro Distinction Water Still, 220 VAC
Supplier:  Cole-Parmer 9929501
Encompass_Preferred
This all-glass constructed water still with silica-sheathed heaters and high-efficiency condenser ensures top distillate purity at an economical price. Special control device automatically turns off the heater to prevent overflow of the reservoir vessel even if the still is left on when unattended. Two independent safety thermostats, mounted in the condenser and boiler, prevent overheating in the event the water supply is interrupted. Screw thread connections make it easy to change hoses quickly and safely without the risk of breaking any glassware. Built-in acid addition funnel and drain stopcock allow for easy descaling without dismantling the glassware. The benchtop still can also be wall mounted using the two predrilled screw holes in the stand.
Catalog No. 50-196-0455