Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cellucap Water Repellent SMS Coats
$123.00 - $283.00
Specifications
Color | White |
---|---|
Closure Type | Snaps |
For Use With (Application) | For dusty areas and superb for splash protection |
Catalog Number | Mfr. No. | Construction | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Construction | Size | Price | Quantity & Availability | ||||
19-022-285
|
Cellucap
3302XXL |
1.25 oz. Spunbond Polypropylene | 2X-Large |
Case of 25 for $123.00
|
|
19-168-280D
|
Cellucap
3503XL |
2-ply Spunbond Polypropylene, inner layer of Meltblown | X-Large |
Case of 25 for $211.00
|
|
19-042-099
|
Cellucap
434TCX |
Polycoated Polypropylene | X-Large |
Case of 50 for $283.00
|
|
19-168-280F
|
Cellucap
35033X |
Spunbond Polypropylene, Meltblown | 3X-Large |
Case of 25 for $227.00
|
|
19-168-280E
|
Cellucap
35032X |
2-ply Spunbond Polypropylene, inner layer of Meltblown | 2X-Large |
Case of 25 for $216.00
|
|
19-168-280C
|
Cellucap
3503LG |
2-ply Spunbond Polypropylene, inner layer of Meltblown | Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Specifications
White | |
Snaps | |
For dusty areas and superb for splash protection |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title