Learn More
Cole-Parmer Thermo Scientific 6140-0010 Faucet Aspirator/Vacuum Pump; 28.5"Hg
Supplier:  Cole-Parmer 0637830
Encompass_Preferred
Perfect for basic laboratory vacuum needs, such as aspiration and filtration. System tubing connection is a detachable stepped hose barb that accepts 1/4" to 3/8" inner diameter flexible tubing. Integral check valve will also prevent backflow of water into the vacuum line.
Catalog No. NC9762985