Learn More
FUJIFILM IRVINE SCIENTIFIC INC Water for Injection, 20 L
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 930920LC
Encompass
Water for Injection (WFI)* Quality Water is the most suitable water available for producing cell culture media, reconstitution of biochemical reagents, and as a final rinse water for critical applications. WFI Quality Water meets USP, PhEur, and JP grade specifications and is 0.1 um sterile filtered at time of fill. It contains no added substances.
Catalog No. NC1328727
Due to product restrictions, please to purchase or view availability for this product.