Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, Deionized, Honeywell™
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
UN Number | NONH for all modes of transport |
Beilstein | 2050024 |
Formula Weight | 18.02g/mol |
Packaging | Poly Bottle |
Density | 1.000g/mL at 3.98°C |
Quantity | 1 L |
Chemical Name or Material | Water |
Safety and Handling
RTECSNumber | 231-791-2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction