Learn More
United Scientific™ Water Cycle Activity Kit
This United Scientific™ Water Cycle Activity Kit demonstrates how water moves through various stages on the earth’s surface. An activity guide is included.
Supplier: United Scientific™ UNWTRCYC02
Alert:
Prop 65 Warning: Cancer and reproductive harm
Description
This United Scientific™ Water Cycle Activity Kit demonstrates how water moves through various stages on the earth's surface. Students will explore the different processes of the water cycle.
- An incandescent lamp (user-provided) simulates the sun, supplying heat to evaporate water in the reservoir below
- Ice cubes are used to cool the water vapor, causing water droplets to form on the cloud
- The water falls onto the mountains, then flows back down to the reservoir
- Activity guide included
- Ten 8.5 x 11 in. full color study cards will enhance the students understanding by showing real life landscapes from the water cycle
Specifications
5 to 12 | |
English | |
Geology |