Learn More
Zymo Research Corporation Water Concentrating Buffer, 8 ml
Supplier: Zymo Research Corporation R204218
Water Concentrating Buffer allows for the effective concentration of microbes, fungi, and viral particles. Especially helpful to concentrate larger volumes up to 5 mL of water by adding Water Concentrating Buffer.