Learn More
Heidolph™ Water Channel for Laborota 20, Hei-VAP Industrial Rotary Evaporator
Water channel collects condensate water and flows it back to the heating bath.
Supplier: Heidolph™ 036280123
Description
Included with Laborota 20, Hei-VAP Industrial Rotary EvaporatorSpecifications
Accessory | |
Laborota 20, Hei-VAP Industrial Rotary Evaporator |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov