Learn More
Chemglass Life Sciences 17L Water Bath, Shaking, Reciprocating, 120V
SDP
Supplier:  Chemglass Life Sciences CLS4952017
Encompass_Preferred
17L Water Bath, Shaking, Reciprocating, 120V. The CLS-4952 Series Reciprocating Water Baths deliver precise temperature control & a smooth reciprocal shaking motion. Temperature & oscillation controls feature an easy to read P.I.D. microprocessor temperature controller & an independent safety thermostat. The bath may be used with or without oscillation. Shaking speed is variable from 20 to 200 oscillations per minute, & the stroke length can be adjusted to three separate settings: 0.5, 1.0, or 1.5 inches to minimize the chance of accidental spillage. The unique basket support system is designed to eliminate tank wear, yet still provide full support & free action for a heavily loaded basket. A durable finish on the housing prevents corrosion & chipping. The bath platform tray is designed for optimum temperature uniformity. Max temperature is achieved using the stainless steel gable cover, designed to prevent condensation from dripping onto samples.
Catalog No. 50-143-555
Explore available promotions