Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water 98% and Acetonitrile 2% with 0.1% TFA, LabReady™ Solvent Blend, for HPLC, Burdick & Jackson, Honeywell
Click to view available options
Quantity:
4 x 4 L
Description
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Max % | 0.10, 1.60, 98.30 |
Density | 0.9820 g/cm3 |
Quantity | 4 x 4 L |
Flash Point | 56.6°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Grade | HPLC |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | H370+H226 |
Precautionary Statement | P210+P233+P240+P241+P242+P243+P260+P264+P270+P280 |
Recommended Storage | Room Temperature |
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction