Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water with 0.1% trifluoroacetic acid, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS, LabReady™ Solvent Blend
$744.00
Specifications
Description | For Use With: LC-MS, Contains 0.093-0.107 % (w/w) trifluoroacetic acid as additive |
---|---|
For Use With (Application) | LC-MS |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
60-090-587
|
Honeywell Chemicals
349784X4L |
4 x 4 L |
Case of 4 for $744.00
|
|
|||||
60-090-586
|
Honeywell Chemicals
349782.5L |
2.5 L | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
For Use With: LC-MS, Contains 0.093-0.107 % (w/w) trifluoroacetic acid as additive | |
LC-MS |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title