Learn More
SCP Science Watch Glass, 50ml, Disposable, 1000 per Box
Supplier: SCP Science 010500081
Unique ribbed watch glasses are designed to fit DigiTUBEs and Teflon tubes to help promote gentle refluxing. Manufactured using virgin polypropylene with ultra low metal content. 50mL. Packaging: Dispenser box