Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ ISOSTAT™ Consumables
For use with ISOLATOR™ 10mL Microbial Tubes.
Supplier: Abbott™ 50C9
Description
The ISOSTAT™ Consumables are used to process the ISOLATOR™ 10mL Microbial Tubes. The ISOLATOR™ 10 mL adult tube is used to concentrate and deliver microbial organisms to conventional growth media for purposes of isolation, identification and susceptibility testing. Show more
- Specifications
- Cap: A plastic cap which penetrates the top stopper of the ISOLATOR™ 10mL tube and permits access to the tube's contents through a membrane-covered port
- Supernatant pipet: A plastic pipet used for removing supernatant fluid from ISOLATOR™ microbial tubes
- Concentrate pipet: Plastic pipet for removing concentrate from ISOLATOR™ microbial tubes to agar plates
Specifications
Isostat Consumables Package | |
Sterile | |
ISOLATOR™ 10 mL Microbial Tubes | |
100 Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction