Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Walter Stern Scalpel Blade Series 300
Fits handles No.3, 3L, 4, 4L and 7
$149.00 - $162.90
Specifications
Blade Material | Stainless Steel |
---|---|
Product Type | Scalpel Blade |
Sterility | Sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
13-812-52
|
Walter Stern
320002 |
# 11 |
Pack of 100 for $149.00
|
|
|||||
17-654-456
|
Walter Stern
320001 |
# 10 |
Pack of 100 for $162.90
|
|
|||||
Description
- Made of Stainless-steel
- Sterile and Individual sealed
Specifications
Stainless Steel | |
Scalpel Blade | |
Sterile |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title