Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Walter Stern Porcelain 6mm Berl Saddles for Distillation Columns
Ideal for vacuum distillation, absorption, extraction
Supplier: Walter Stern 500A
Description
- Relatively high usable contact surface area Lower gas flow resistance Good internal distribution
Order Info
In 200cc packs.
Specifications
For vacuum distillation, absorption, extraction, can also be used as boiling stones | |
Porcelain | |
For distillation columns |
Berl Saddles | |
120cm3 | |
7.32 in.3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title