Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Walter Products 2-Stage Vacuum Pump
Portable, ready to operate vacuum pump.
Supplier: Walter Products P30001
Description
- Easy-to-carry design
- Provides free air displacement of 2.5CFM at 50Hz and 3.0CFM at 60Hz
- Runs steadily for long period of time
- Motor: 1/3HP
- Intake Ports: 6.35mm (0.25 in.)
Specifications
3 x 10-1 Pa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction