Learn More
Mopec™ Waffle Specimen Chucks for SHANDON Cryostats
Waffle Specimen Chucks are available in multiple sizes and colors.
Supplier: Mopec™ SP144WWH
Description
- Round, aluminum specimen chucks
Specifications
Waffle Chuck | |
SHANDON Cryostats | |
40 mm | |
White |