Learn More
Andwin Scientific VWR STATICMSTR IONIZING UNITV
SDP
Supplier:  Andwin Scientific 58580041/IU
Encompass_Preferred
Ionizer, Balance Chamber, 500 MIC Self-Powered, Plonium-210 Ion Source, mount near Pan on Chamber Floor/Wall, Staticmaster, 3W x 1 in. D
Catalog No. NC9506888
Explore available promotions