Learn More
Andwin Scientific Culture Polypropylene Tubes, 5ml, 12 x 75 mm, 1000/case
Supplier: Andwin Scientific 60818500
The culture polypropylene tubes have round bottoms, volume capacity of 5 mL, and dimension of 12 x 75 mm. The tubes can resist autoclaving temperature up to 122C (252F) and freezing temperature down to -90C (-194F). NC0325964 as non returnable and sterile.