Learn More
Volta's Hailstorm
Create a tempest in a teapot.
Supplier: Science First 6153155
Includes: Transparent cage (12 in. H [31cm]) mounted on sturdy metal base (9 in. dia. [23cm]); ball terminal to attract electric charge; tiny polystyrene balls and instructions with theory
Description
Great for demonstrating Coulomb's law qualitatively and illustrating the method of smoke control based on the electrostatic removal of solid particles from the smoke stream.
- Consists of a transparent cage with a metallic ceiling and floor
- Tiny conductive balls inside begin to bounce when near an active Van de Graaff generator
- Results in what looks like a hailstorm
Specifications
Demonstrating Coulomb's law qualitatively and illustrating the method of smoke control based on the electrostatic removal of solid particles from the smoke stream | |
Transparent cage (12 in. H) mounted on sturdy Metal base (9 in. dia.) ball terminal to attract electric charge, tiny Polystyrene balls and instructions with theory | |
Voltas Hailstorm Electric Demonstration Aid |