Learn More
Restek™ Viva Guard Cartridges
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Particle Size: 5 μm (Spherical)
$256.00 - $327.00
Specifications
Length (Metric) | 10 mm |
---|---|
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 300 Å |
Quantity | 3/Pk. |
Catalog Number | Mfr. No. | Type | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Diameter (Metric) | Price | Quantity & Availability | ||||
06-611-680
![]() |
Restek™
951650212 |
Guard Cartridge | 2.1 mm |
Pack of 3 for $256.00
|
|
||||
06-611-679
![]() |
Restek™
951650210 |
Guard Cartridge | 4 mm |
Pack of 3 for $307.00
|
|
||||
06-611-674
![]() |
Restek™
951250212 |
Guard Cartridge | 2.1 mm |
Pack of 3 for $256.00
|
|
||||
06-611-673
![]() |
Restek™
951250210 |
Guard Cartridge | 4 mm |
Pack of 3 for $289.00
|
|
||||
06-611-676
![]() |
Restek™
951350212 |
Guard Cartridge | 2.1 mm |
Pack of 3 for $256.00
|
|
||||
06-611-675
![]() |
Restek™
951350210 |
Guard Cartridge | 4 mm |
Pack of 3 for $307.00
|
|
||||
06-611-678
![]() |
Restek™
951450212 |
Guard Cartridge | 2.1 mm |
Pack of 3 for $256.00
|
|
||||
06-611-677
![]() |
Restek™
951450210 |
Guard Cartridge | 4 mm |
Pack of 3 for $327.00
|
|
||||
06-611-682
![]() |
Restek™
951950212 |
Guard Cartridge | 2.1 mm |
Pack of 3 for $256.00
|
|
||||
06-611-681
![]() |
Restek™
951950210 |
Guard Cartridge | 4 mm |
Pack of 3 for $307.00
|
|
||||
06-611-672
![]() |
Restek™
951050212 |
Guard Cartridge | 2.1 mm |
Pack of 3 for $271.00
|
|
||||
06-611-671
![]() |
Restek™
951050210 |
Guard Cartridge | 4 mm |
Pack of 3 for $271.00
|
|
||||
Description
- Gaurd Columns require a Trident LC Column Protection System or Trident HPLC In-Line Guard Cartridge Holder
Specifications
10 mm | |
5 μm | |
3/Pk. |
Spherical | |
300 Å | |
Guard Cartridge |