Learn More
Perkin Elmer US LLC Viton Torch Injector O-Ring, 6.07 mm I.D.
Supplier: Perkin Elmer US LLC 09902207
Two Viton 6.07 mm I.D. o-rings are included in the Scott/Cross-Flow Torch Injector O-Ring Kit for Avio 200/500 (N0790440), the Cyclonic/Concentric Torch Injector O-Ring Kit for Avio 200/500 (N0790441), and the Complete O-Ring Kit for Optima 4300V/5300V/7300V (N0770916).