Learn More
CTL Scientific Supply Corp VISOCOLOR ECO PHOSPATE
SDP
Supplier:  CTL Scientific Supply Corp 931084
Encompass_Preferred
Environmental test kit for Phosphate Gradation of scale: 0.2 - 0.3 - 0.5 - 0.7 - 1 - 2 - 3 - 5 mg/l 80 tests/kit
Catalog No. 50-300-382