Learn More
CTL Scientific Supply Corp VISOCOLOR ECO CHLORIDE
SDP
Supplier:  CTL Scientific Supply Corp 931018
Encompass_Preferred
Environmental test kit for Chloride Gradation of scale: 0 - 1 - 2 - 4 - 7- 12 - 20 - 40 - 60 mg/l 90 tests/kit
Catalog No. 50-300-367