Learn More
Eppendorf™ Pipette Manager PROMO
Pipette manager allows you to easily transfer volumes to connected Eppendorf™ electronic pipettes and much more.
Supplier:  Eppendorf™ 2231000910
Catalog No. 02911585PM
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Pipette Manager, an external touch server enabling the communication with connected Eppendorf Xplorer electronic pipettes.
  • Easily enter pipetting volumes and other parameters using the touch panel. Settings are immediately transferred to connected electronic pipettes, allowing you to start working right away
  • Get guidance from the software-embedded Liquid Manager when working with liquids of different viscosity, volatility or foam-building liquids
  • Choose from predefined Eppendorf liquid types (e.g. acetone) or create your own
  • Automatically document pipetting activities via Pipetting Records and easily export your recordings as PDF file via USB
  • Manage your pipette fleet more effectively within your working group and enhance collaboration (e.g. through sharing created liquid types)
  • All Eppendorf Xplorer and Xplorer plus 4, 6, 8 and 12-channel electronic pipettes are compatible. Move It™ pipettes are currently not supported.
  • Easily upgrade Eppendorf Xplorer and Xplorer plus electronic pipettes with the Eppendorf Xplorer connect WiFi module. The WiFi modules comes with shorter battery for an optimal fit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
External Touch Server
Product Suggestions
Product Suggestions