Learn More
Azzota Corp Vis Neutral Density Filter Set Spectrophotometer Cali Tools For Photometric Accuracy Total 3 Filters: 10%, 20% And 30% Check Multi-Wavelength At 400Nm, 546Nm And 750Nm Filter Size 10 X 10 X 45 mm
Supplier:  Azzota Corp Q807
Encompass_Preferred
VIS Neutral Density Filter Set Spectrophotometer calibration tools Use for testing Spectrophotometer Photometric Accuracy Total 3 filters: 10%, 20% and 30% Check Multi-Wavelength at 400nm, 546nm and 750nm Filter size 10 x 10 x 45 mm
Catalog No. 50-753-2878
Explore available promotions