Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio VirusGEN AAV Transfection Kit with RevIT AAV Enhancer

Paired with TransIT VirusGEN, the RevIT AAV Enhancer delivers higher titers for recombinant adeno-associated virus production

$1205.00 - $9104.00

Specifications

Cell Type 293-derived Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Product Type AAV Transfection Kit with Enhancer
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
MIR8007
View Documents
Mirus Bio
MIR 8007
1 L
Each for $1,205.00
Add to Cart
 
MIR8008
View Documents
Mirus Bio
MIR 8008
10 L
Each for $9,104.00
Add to Cart
 
Description

Description

  • TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
  • Kit includes: TransIT-VirusGEN Transfection Reagent and RevIT AAV Enhancer 
  • Key benefits: 
    • Performance: Efficient DNA delivery for production of high-titer viral vectors
    • Scalability: Efficient production from less than 1 mL to large scale culturesFlexibility: Compatible with different recombinant virus and cell culture systems
    • Animal Origin Free: Fully synthetic transfection reagent formulation
Specifications

Specifications

293-derived Cells
None (Promoterless)
AAV Transfection Kit with Enhancer
Tube
Chemical Transfection
Sterile
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit