Learn More
Remco™ Vikan™ Soft Overhead Brush
Remco™ Vikan™ Soft Overhead Brush is designed to sweep away dust and fine particles when cleaning the tops of pipes.
Supplier: Remco™ 53713
Description
- Soft CIP brush
- Great for cleaning overhead pipes when used with a telescopic handle
- Dirt falling from overhead equipment is a high risk
- Soft bristles help clean off dust-like particles from hard-to-reach spots
- The neck is adjustable to be used at different angles
- Adjust the angled head to provide a custom cleaning experience for any processing facility
- Color-coded
Specifications
Pipe Brush | |
Medium Stiffness | |
Stainless Steel | |
Blue | |
1 Ea. |
Polyester | |
Yes | |
Non-sterile | |
21.7 in. | |
Medium |