Learn More
Remco™ Vikan™ Small Utility Brush
Remco™ Vikan™ Small Utility Brush is the ideal cleaning tool due to its size, ergonomically friendly shape, and versatile bristles.
Supplier: Remco™ 30886
Description
- Broad head
- Ergonomically angled handle
- This design raises the user's hand from the cleaning surface reducing the risk of injury
- Suitable for cleaning small surface areas, tables, pots, cutting boards, and more
- Color-coded
Specifications
Utility Brush | |
Yes | |
Non-sterile | |
Yellow | |
1 Ea. |
Polyester | |
Polypropylene | |
FDA Compliant | |
10.24 in. | |
Small |