Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Vikan™ Short Handle Utility Brush
Remco™ Vikan™ Short Handle Utility Brush is a short-handled washing brush with medium bristles.
$18.00
Specifications
Product Type | Utility Brush |
---|---|
Bristle Material | Polyester |
Bristle Type | Medium Stiffness |
Color-coded | Yes |
Handle Material | Polypropylene |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
17-227-288
|
Remco™
41904 |
Red |
Each for $18.00
|
|
|||||
17-227-290
|
Remco™
41906 |
Yellow | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
17-227-286
|
Remco™
41902 |
Green | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
17-227-287
|
Remco™
41903 |
Blue | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
17-227-289
|
Remco™
41905 |
White |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
- Comfortable grip
- Allows you to work for long periods of time without hand fatigue
- Medium strength bristles are great for washing all types of equipment, tables, and walls
- Brush can be used holding both the handle and the brush head for better control
- Color-coded
Specifications
Utility Brush | |
Medium Stiffness | |
Polypropylene | |
FDA Compliant | |
1 Ea. |
Polyester | |
Yes | |
Non-sterile | |
9.8 in. | |
Short |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title