Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Vikan™ Medium Tank Brush
Remco™ Vikan™ Medium Tank Brush is ideal for cleaning tanks, vats, and drain gulleys.
$32.55 - $33.60
Specifications
Product Type | Cleaning Brush |
---|---|
Bristle Material | Polyester |
Bristle Type | Medium Stiffness |
Color-coded | Yes |
Handle Material | Polypropylene |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
17-227-765
|
Remco™
70394 |
Red |
Each for $32.55
|
|
17-227-767
|
Remco™
70396 |
Yellow |
Each for $33.60
|
|
19-830-209
|
Remco™
70392 |
Green |
Each for $33.60
|
|
17-227-764
|
Remco™
70393 |
Blue |
Each for $33.60
|
|
17-227-766
|
Remco™
70395 |
White |
Each for $33.60
|
![]() |
Description
- Ideal for cleaning tanks and vats
- Suitable for cleaning drain gulleys
- Can be used with any European-thread handle from Vikan™
- Color-coded
Specifications
Cleaning Brush | |
Medium Stiffness | |
Polypropylene | |
FDA Compliant | |
8.3 in. | |
Large |
Polyester | |
Yes | |
Non-sterile | |
Thread type Euro Hygienic | |
1 Ea. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title