Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ Medium 4 in. Diameter Pipe Brush

Remco™ Vikan™ Medium 4 in. Diameter Pipe Brush can be used dry to sweep off tough dirt, or for scrubbing when combined with water and a cleaning agent.

Supplier:  Remco™ 53801039

 View more versions of this product

Catalog No. 17-227-428


Add to Cart

Description

Description

  • Features bristles along the sides and in the front
  • It is used in areas where a wider tube cleaning action is required or where the equipment has an endpoint that cannot be reached with a hand brush
  • Works well as a drain brush
  • Requires a European-thread handle
  • Color-coded
Specifications

Specifications

Pipe Brush
Medium Stiffness
Polypropylene
FDA Compliant
4 in.
1 Ea.
Polyester
Yes
Non-sterile
Black
6.5 in.
Medium
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit