Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Vikan™ Lid for 3 Gallon Bucket
Remco™ Vikan™ Lid for 3 Gallon Bucket has a clip system for keeping the lid in place when in use.
Supplier: Remco™ 56878
Description
- Ideal for preventing spillage of materials
- It has a clip system for securing the lid tightly
- For use with 3 Gallon Bucket
- Color-coded
Specifications
Not Autoclavable |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction