Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Vikan™ Color-Coded Stainless Steel Floor Scrapers
Remco™ Vikan™ Color-Coded Stainless Steel Floor Scrapers can be combined with any Euro thread handle to get to hard-to-reach areas.
Supplier: Remco™ 29106
Description
- Ideal for removing stubborn debris and other substances
- Long-lasting blade
- Connects with any Remco™/Vikan™ handles to provide an easy solution for stuck-on messes
- Scapper is hygienically designed for easy cleaning
Specifications
Floor Scraper | |
Yellow | |
Cleaning hard-to-reach areas | |
1 Ea. | |
0.56 lb. |
FDA Compliant | |
10.25 x 1.25 in. | |
Polypropylene, Stainless Steel | |
Large |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction