Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ Color-Coded Basic Wall Bracket

Remco™ Vikan™ Color-Coded Basic Wall Bracket is a one-piece wall bracket with mounting kit that can hold up to three products.

$34.95

Specifications

Type Wall Bracket
Material Polypropylene
For Use With (Application) Color-coded storage
Certifications/Compliance FDA Compliant
Quantity 1
View More Specs

Products 7
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
17-227-003
View Documents
Remco™
06154
Red
Each for $34.95
Add to Cart
 
17-227-006
View Documents
Remco™
06157
Orange
Each for $34.95
Add to Cart
 
17-227-005
View Documents
Remco™
06156
Yellow
Each for $34.95
Add to Cart
 
17-227-001
View Documents
Remco™
06152
Green
Each for $34.95
Add to Cart
 
17-227-002
View Documents
Remco™
06153
Blue
Each for $34.95
Add to Cart
 
17-227-008
View Documents
Remco™
06159
Black
Each for $34.95
Add to Cart
 
17-227-004
missing translation for 'viewDocuments'
Remco™
06155
White
Each for $34.95
Add to Cart
 
Description

Description

  • Supports any product and is virtually unbreakable
  • Includes screws, wall anchors, and mounting instructions
  • Provides a color-coded storage option for tools
  • Proper storage of cleaning equipment
  • Supports hygienic environments
  • Helps to extend the usable life of the equipment
Specifications

Specifications

Wall Bracket
Color-coded storage
1
Polypropylene
FDA Compliant
9.4 x 6.5 x 2.25 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit