Learn More
Beckman Coulter VIAL SCINT NO LINER 1000/CS
Supplier: Beckman Coulter 566740
POLY-Q Vials bulk pack of 1 000 vials w/ caps 18mL POLY-Q vials are made of high density linear polyethylene and have a very low permeability for the organic solvents used in scintillation cocktails