Learn More
Beckman Coulter FisherbrandTM Petri Dishes, 100 x 15 mm, Sterile, 500/Pk
Supplier: Beckman Coulter 90620008
The FisherbrandTM NC2281877 is a polypropylene microcentrifuge tube with a snap cap. It has a capacity of 1.5 mL and dimensions of 10.8mm x 38mm. The tubes are made of clear polypropylene for visibility. They can withstand centrifugal forces up to 20,800 x g and temperatures from -86°C to 121°C. The attached snap caps form a tight seal and the ribs prevent the tube from rolling. These microcentrifuge tubes are sterile, free of DNase, RNase and pyrogens and come packaged in bags of 500.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.