Learn More
Hach Company Vial, Precleaned Glass, 40 mL, with cap, pk/5
Supplier: Hach Company 2794005
Capped clear glass vial precleaned to USEPA standards for volatile organics analyses. 40 mL size is good for collecting TOC samples for Hach TOC tests.