Learn More
TWD Scientific LLC BioX Tricode 2.0mL Cryovial, Purple Cap, Internal Thread. Case of 1000
Supplier: TWD Scientific LLC BX889215
BioX 2.0mL clear polypropylene sterile pre-set 1D & 2D barcoded cryovials with internal thread and purple caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case
This item is not returnable.
View return policy