Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ VHB™ Tape
3M Company VHB™ Tape is a fast and easy-to-use permanent bonding method that provides high strength and long-term durability
Supplier: 3M™ 7000028996
Description
- Virtually invisible fastening keeps surfaces smooth
- Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
- Gray, 0.090 in (2.3 mm), multi-purpose adhesive and conformable acrylic foam core offers a good balance of strength and conformability
- Eliminate drilling, grinding, refinishing, screwing, welding and associated clean-up
- Creates a permanent seal against water, moisture and more
- Multi-purpose
- Conformable foam core
- Can replace rivets, welds and screws
- Offers design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces
- 0.5 in x 36 yd 91.0 mil
- 3M Catalog# 4991
Specifications
Permanent | |
Foam | |
4991 | |
91 mil | |
32.9 m |
Gray | |
Exterior Metal Skin Bonding | |
18/Pk. | |
36 yd. x 0.5 in. | |
12.70 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction