Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Vesta Syde™ SQ 128 st RTU Disinfectant

Sterile version of phosphate-free one-step cleaner and disinfectant, formulated for use on hard, non-porous surfaces

$842.50 - $943.50

Specifications

Product Type Quaternary Disinfectant
pH 10.4
Sterility Sterile
Composition Quaternary Ammonium
View More Specs

Products 3
Catalog Number Mfr. No. Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Price Quantity & Availability  
17-463-008
View Documents
STERIS™
1S25Y4
16 oz.
Case for $853.00
Add to Cart
 
17-463-030
View Documents
STERIS™
1S25Y2
23 oz.
Case of 12 for $943.50
Add to Cart
 
17-463-031
View Documents
STERIS™
1S2508
128 oz. (1 gal.)
Case of 4 for $842.50
Add to Cart
 
Description

Description

  • Kills broad spectrum of common bacteria and fungi
  • Use-dilution rate of 0.8% v/v (1:128)
  • Requires no mixing or activation for convenient and consistent use
  • Specifically formulated for disinfection of hard surfaces
  • Can be applied manually or sprayed
  • Ease of Use- Eliminates need to validate dilution preparation procedures by providing consistent dilution every time
  • Convenience- Same key ingredients as Vesta-Syde™ SQ Disinfectant concentrate to simplify validation requirements
  • Productivity- Low residue formulation to maintain visibly clean surfaces with minimal rinsing
  • Efficacy- Microbial control against bacteria, fungi, and viruses using AOAC and EN protocols
  • Availability- Active ingredients notified according to Biocide Product Directive and registered with US EPA
  • Assurance- Manufactured in accordance with cGMP with complete lot traceability
Specifications

Specifications

Quaternary Disinfectant
10.4
Sterile
Quaternary Ammonium
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit