Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Steris™ Vesta-Syde™ SQ Ready-to-Use Disinfectant
Vesta-Syde™ SQ Ready-to-Use (RTU) Disinfectant is a phosphate-free, one-step cleaner and disinfectant formulated in two concentrations for use on hard, non-porous surfaces.
Supplier: Steris™ 634308
This item is not returnable.
View return policy
Description
- Easy to use
- Saves the time and labor of disinfectant preparation
- Eliminates errors in dilution preparation and the need to validate use procedures
- Confirms efficacy against many common microbes
- Simplifies validation requirements and offers multiple options for preparation
- Ability to maintain visibly clean facilities with minimal rinsing
- Allows for high frequency use on most surfaces
- Available in most countries worldwide to facilitate program harmonization
- Allows easy entry into controlled environments
- Supports quality assurance and quality control goals
- Formulation is ready to use without any mixing
- Broad spectrum efficacy against bacteria, fungi, and viruses using AOAC and EN protocols
- Same key ingredients as Vesta-Syde SQ Disinfectant concentrate
- Low residue formulation that is easily rinsed if required
- Superior material compatibility
- Active ingredients notified according to the Biocide
- Product Directive and registered with the US EPA
- Manufactured in accordance with cGMP with complete lot traceability
- Designed for use on:
- Countertops
- BSL hoods
- Carts and other equipment
- Floors
- Walls
- Ceilings
- Products may be applied by:
- Mopping
- Spraying
- Wiping
Specifications
Non-sterile | |
Filtered | |
3.78 L |
0.8% v/v (1:128) | |
1 gal. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction