Learn More
Chemglass Life Sciences 10mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 14 Thread
SDP
Supplier:  Chemglass Life Sciences CG186110
Encompass_Preferred
10mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 14 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D.
Catalog No. 50-153-1263