Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC VERTREL XF CLEANING AGENT 5GAL
Supplier: MILLER STEPHENSON CHEMICAL COMPANY INC MS7825GAL
Cleaning Agent, Vertrel; XF, A Very Mild, Nonflammable Solvent. Excellent Compatibility with Most Plastics. Typical Applications Include: Cleaning, Rinsing, Drying, Particulate Removal, Fluorocarbon Lubricant Carrier, Heat Transfer, 5 gal.